Print…

Anti-MYRIP Goat Polyclonal Antibody

Fornecedor: OriGene
ORIGTA302880EA 547 EUR
ORIGTA302880
Anti-MYRIP Goat Polyclonal Antibody
Anticorpos
Recommended Dilutions: ELISA: 1:64000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: MYRIP (myosin VIIA and Rab interacting protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR