Print…

Anti-MESP1 Rabbit Polyclonal Antibody

Fornecedor: Aviva Systems Biology
AVIVARP39374_P050EA 371 EUR
AVIVARP39374_P050
Anti-MESP1 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: MESP1 (mesoderm posterior 1 homolog (mouse))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR