Print…
Magnetic stirrer, mini, SM5
Fornecedor: Cole Parmer
Stuart®
STUASM5/SWIRLEA 246 EUR
STUASM5/SWIRL STUASM5/BIBBY
Magnetic stirrer, mini, SM5
Agitadores magnéticos e suspensos Agitadores magnéticos
The SM5 mini stirrer is a powerful magnetic stirrer, capable of stirring up to 1 l, the stirring speed is adjustable up to 2000 rpm. The mini stirrer is available in a choice of designs. The unit is made from robust polypropylene with a chemically resistant polycarbonate top.
  • Lightweight stirrer
  • Universal power adaptor
  • With BioCote antimicrobial protection