Print…
Volumetric flasks, class A, with screw cap, PYREX®
Fornecedor: Corning
PYREX®
CORN5650-50EA 0 EUR
CORN5650-25 CORN5650-100 CORN5650-50
Volumetric flasks, class A, with screw cap, PYREX®
Balões Balões Volumétricos
Made of PYREX® borosilicate glass.
Flasks have an expanded open end to facilitate transfer of crystals, salts and powders. Standard GPI thread screw cap has an inert, cone-shaped, polyethylene liner for leakproof closure. Conforms to Class A capacity tolerances.