Print…
pH meter modules for for iPhone®, iPod® & iPad®
VWR®
662-1489EA 277 EUR
VWRI662-1489
pH meter modules for for iPhone®, iPod® & iPad®
This device measures and records pH values in the lab or field using the iPhone®, iPod® and iPad®. The supplied app provides real time display of pH, mV, ambient temperature and solution temperature. Simply plug in the PH-1 to your iPhone®, iPod® and iPad® and follow instructions to download the free app from the App Store. The PH-1 requires operating system OS 4.0 or higher.
  • When used with a GPS enabled device, the pH meter app will record where and when measurements were taken
  • Results can be grouped by location or date, and emailed to a central records office
  • Ideal for the classroom: Studets can install the app on their own iPod® or iPhone®, and share a single sensor for experiments
  • Suitable for any process where pH measurements are routinely performed
  • Displays both ambient and solution temperature
  • Supports 1, 2, 3 or more calibration points
Connecting instructions: IOS4 and 5 Users – Simply attach the PH-1 to your device. You should be automatically prompted to download the free app. IOS6 users – Attach the PH-1 to your device. Go to the app store and search for pH Meter. Download the free app. Newer devices with the lightning connector will need to use a 30 pin adapter which can be sourced from Apple™.
Apple™, iPhone®, iPod® and iPad® are registered trademarks of Apple Inc.
Informações de encomenda: pH meter module is supplied without electrode (needs to be ordered separately). iPod®, iPhone® or iPad® not included.