Print…
Conductivity meter, handheld, HQd (HQ14d)
Fornecedor: Hach
HACHHQ14D99201000EA 898 EUR
HACHHQ14D53000000 HACHHQ14D99201000 HACHHQ14D99203000
Conductivity meter, handheld, HQd (HQ14d)
This portable conductivity meter combines reliability, flexibility and ease of use. Interchangeable INTELLICAL™ electrodes are recognised automatically and store all relevant data.
  • Rugged, waterproof (IP 67) meter provides worry-free, reliable operation in lab or field environments
  • Guided calibration and check standard routines reduce calibration errors
  • Stabilisation alerts and visual measurement lock ensure that you can trust the accuracy of the results
  • Full GLP data management
Ideal for measuring critical water quality parameters.
Fornecimento: Meter only (without electrode) is supplied with 4 AA batteries, quick-start guide, and user manual. The kits additionally include conductivity cells and reagents.