This portable conductivity meter combines reliability, flexibility and ease of use. Interchangeable INTELLICAL™ electrodes are recognised automatically and store all relevant data.
- Rugged, waterproof (IP 67) meter provides worry-free, reliable operation in lab or field environments
- Guided calibration and check standard routines reduce calibration errors
- Stabilisation alerts and visual measurement lock ensure that you can trust the accuracy of the results
- Full GLP data management
Ideal for measuring critical water quality parameters.
Fornecimento: Meter only (without electrode) is supplied with 4 AA batteries, quick-start guide, and user manual. The kits additionally include conductivity cells and reagents.