Print…
Column connectors, Multifit
Fornecedor: Spectrum
SPEC124487EA 93.8 EUR
SPEC124487
Column connectors, Multifit
The Multifit connectors are used for connecting flexible tubing to the column end-plates. The aqueous series connectors are available with an O-ring for gripping the tubing and the organic series connectors are available with a PTFE ferrule.