Print…
Vacuum pipetting device, Vadosa
Fornecedor: Kartell
KART199EA 33.4 EUR
KART199
Vacuum pipetting device, Vadosa
Pipetting aid for pipettes from 0,1 ml up to 25 ml.
  • Will deliver even the smallest quantities with simplicity, precision and speed
  • Simple, one handed operation with no hand squeezing, no sucking, no guessing
  • Made of acid and alkali-resistant PP
This safety vacuum pipetting device is easy to use, being able to draw up and measure any liquid, even the most viscous ones. It fits every type and size of pipette from 0,1 ml up to 25 ml.