Print…
Tubing connectors, stepped, SP Bel-Art
Fornecedor: Bel-Art Products, a part of SP
BELAH195630000EA 69.2 EUR
BELAH195630000
Tubing connectors, stepped, SP Bel-Art
Conexões e Conectores
PP, translucent.
  • Excellent chemical resistance for use with a wide range of liquids or gases
  • Excellent temperature resistance, autoclavable
  • FDA-grade for direct contact with food, drugs, and cosmetics
These fittings ensure a tight, secure connection between tubing having different inner diameters (Int. Ø.).