Print…
Sharps disposal containers Multi-Safe opti
Fornecedor: Sarstedt
SARS77.3895.007EA 174 EUR
SARS77.3895.007
Sharps disposal containers Multi-Safe opti
Boxes, made of PP, yellow/red. With a large deposit slot for the disposal of scalpels, used cannulas, disposable syringes, tubing, broken glass and long-stemmed objects.
  • Boxes can be fixed with an adhesive or a wall holder to prevent them from overturning (see accessories), Exception model 500T
  • Universal stripping/detaching devices also for insulin pens
Certificações: TRBA 250, UN, DIN EN ISO 23907