Print…
Adapters, solvent still head, Quickfit®
Fornecedor: SciLabware
BIBBSH9/1LEA 1790 EUR
BIBBSH9/1L BIBBSH9/250
Adapters, solvent still head, Quickfit®
PYREX®, borosilicate glass.
  • Solvent removed from the flask via top stopcock by syringe or drainage through T-bore stopcock
  • Drainage arm utilises a 14/23 cone
  • Stopcocks with PTFE key
Apparatus for distillation and collection of oxygen-free solvents.