Print…
Microcentrifuge tubes
Fornecedor: Biotix
525-0580EA 23 EUR
525-0580
Microcentrifuge tubes
Tubos Tubos de microcentrífuga
PP, graduated, autoclavable and freezable with attached flat caps.
  • DNase-, RNase and endotoxin-free
  • Can withstand centrifugation to RCF 20000×g
  • Tubes with X-Resin™ provides ultra-low binding characteristics
Medical grade superior clarity for visualisation of samples and isolated pellets. Graduated frosted side panel and flat snap cap provide surface for labelling.
Assorted colours pack includes red, blue, green, yellow, orange tubes