Print…
Emergency body and eye showers, for wall mounting
Fornecedor: Broen Lab
BROE17.452.009EA 1430 EUR
BROE17.452.009 BROE17.455.009
Emergency body and eye showers, for wall mounting
Emergency body and eye shower for wall mounting. Available with and without bowl for eye/face shower. In brass with chemical resistant red polycoating.
  • With self-draining shower head
  • Eye wash with built-in FLOWFIX for regulation of water flow (26 l/min)
  • Water inlet at the top or bottom
  • Pre-mounted valve for body shower
  • Easy to mount and install
Depth: 685 mm
Weight: ~8 kg
Connection: Inlet G3/4" female
Min. working pressure: 1,5 bar
Certificações: EN 15154-1/-2; ANSI Z358.1. TÜV certified