Print…
DAB substrato, tabletes (5mg/240mg) para biotecnologia
Fornecedor: Avantor
VWR®
E733-100TABSEA 164 EUR
E733-100TABS E733-50TABS
DAB substrato, tabletes (5mg/240mg) para biotecnologia
Electroforese, Reagentes Western Blotting Reagents
DAB is commonly used for detection of horseradish peroxidase in membrane-bound and in situ applications.
Insoluble end product is resistant to alcohol and fading, making it ideal for immunohistochemistry. Each tablet contains five mg of DAB and will yield 10 ml of substrate solution, enough to effectively detect 50 cm² of membrane or about 20 slides.
pH (one tab/10 ml water): 4,8 to 5,2.
Embalagem: DAB substrate available in glass bottle.