Print…
Eye and face shower, for in-wall mounting
Fornecedor: Broen Lab
118-1934EA 518 EUR
BROE17.933.409
Eye and face shower, for in-wall mounting
Powder coated brass eye shower for fixed wall assembly.
  • For exposed pipe work
  • Water amount is regulated by a built-in FLOWFIX (26 l/min)
  • Doesn't close automatically
  • Rubber protection with dust cap
  • Plastic plate with clear "PUSH" marking
  • Push-on rosette made from polyamide
  • Stainless steel drip tray (Ø 275 mm) with drain fitting
Connection: G ½"A
Weight: ~4,5 kg
Min. working pressure: 1,5 bar
Certificações: EN 15154-2; ANSI Z358.1. TÜV certified