Print…
Pipette stand, rotary, Azlon®
Fornecedor: SciLabware
Azlon®
AZLOSWP033EA 223 EUR
AZLOSWP033
Pipette stand, rotary, Azlon®
Suportes de pipetas
Rotary pipette stand, made of blue PP.
  • Chemically resistant
  • Holds up to 94 pipettes vertically
  • Very stable with 230 mm diameter base
  • Has a rotatable rack suspended by a support rod