Print…
Sponges, PolyMesh™
Fornecedor: Micronova
115-1816EA 13.7 EUR
115-1816
Sponges, PolyMesh™
Toalhetes Esponjas
Sponge made of reticulated urethane foam with an abrasive polyester mesh outer layer that stands up to heavy soils without shedding or flaking. Ideal for removing chemical deposits, photo resist and fluxes.
  • Autoclavable
  • Resistant to strong disinfectants and steam sterilisation
  • Clean alternative to household grade abrasive pads
  • Ideal for scrubbing heavy soils or dried residues