Cholesterol butyl carbonate ≥95.0% (by HPLC)
Fornecedor: TCI EUROPE
TCIAC0718-10GEA
115
EUR
TCIAC0718-10G
Cholesterol butyl carbonate ≥95.0% (by HPLC)
Cholesterol butyl carbonate
Fórmula:
C₃₂H₅₄O₃ |
Número MDL :
MFCD00059502 Número CAS: 41371-14-6 |
Especificações dos Resultados do Teste
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Mesomorphic range | 69.0 to 93.0 deg-C |
Solubility in hot Acetone | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...