Cholesterol isobutyl carbonate
Fornecedor: TCI EUROPE
TCIAC0719-10GEA
186
EUR
TCIAC0719-10G
Cholesterol isobutyl carbonate
Cholesterol isobutyl carbonate
Fórmula:
C₃₂H₅₄O₃ |
Número MDL :
MFCD00059501 Número CAS: 77546-35-1 |
Especificações dos Resultados do Teste
Appearance | White to Almost white powder to crystal |
Specific rotation [a]20/D | -29.0 to -37.0 deg (C=2,chloroform) |
Solubility in Chloroform | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...