Designed to perform non-pathogenic handlings, the new range of CaptairBio PCR enclosures has been created for handlings involving a high sample cross contamination risk such as sample preparations before thermocycling, Post PCR DNA sequencing revelation / separation, in vitro fertilization, cellular cultures, vegetal biology, preparation of sterile solutions.
- The worksurface made of stainless steel makes the cleaning easy
- Total openings for an easy access into the inclosure
- Energy efficient
- Automatic UV cut-off in case of front door opening
- Internal reflectors: no shadow area
- Serial prefilter optimizes the HEPA H14 filtration
New enclosure design - from 80 cm to 1,8 m –equipped with a powerful UV decontamination preventing any handling from cross contamination risks.
The design of the new modular filtration column equipped with an HEPA H14 filter guarantees 99,995% filtration efficiency for particles larger than 0,1 microns (according to the EN1822-1 standard, MPPS method).
The air entering the enclosure is free of any external contamination and provides an ultra clean work atmosphere.
The modular filtration column technology also allows the use of optional carbon molecular filter offering an additional protection of the enclosure from gase pollutants present in the laboratory ambient air which may interfere with sample integrity.
With their very low energy consumption from 70W to 150W, the annual running cost of these units does remains very low and allows a faster return on investment
Work surface in stainless steel 304 L with built-in retention tray or tempered glass work surface with seeled frame
Note: Please note that the PCR workstations 321 S models are not equiped with filtration column (no HEPA filter).
Fornecimento: Delivery prices, installation and commissioning are available on request.