Print…
Earmuffs, Leightning® L2
Fornecedor: Honeywell Safety
111-1406EA 0 EUR
DALL1011997
Earmuffs, Leightning® L2
Robust and durable earmuffs for rough industrial environments.
  • Medium profile cups
  • Air Flow Control™ Technology provides better, consistent attenuation, especially at low frequencies
  • Snap in ear cushions makes cushion replacement quick and easy
  • Padded foam headband delivers long wearing comfort and reduces the pressure on the head
  • Telescopic height adjustment remains fixed during use
The earmuffs work especially well in extreme temperatures/humidity and under aggressive chemical influences.
SNR: 31 dB (headband)
SNR: 32 dB (foldable)
Weight: 231 g (headband); 236 g (foldable)
Certificações: EN 352-1