Print…
VWR®, Protein EZ-Vision®, 4X Biotechnology grade
Fornecedor: Avantor
VWR®
N836-KITEA 131 EUR
N836-KIT
VWR®, Protein EZ-Vision®, 4X Biotechnology grade
Corantes Corantes para Electroforese
  • Biotechnology grade convenient loading buffer and visualisation dye for proteins
  • Sensitivity for immediate visualisation after electrophoresis
  • Compatible with downstream Western blotting
Loading buffer with fluorescent protein dye offers a non hazardous alternative to Coomassie® Blue staining.
Specifications
Abs.@436nm (1:100, Water): 0.2 - 0.4
Abs.@560nm (1:100, Water): 0.65 - 0.96
Protease: NONE / Fail
Informações de encomenda: Includes two 1 ml tubes.
Atenção: For research use only – not for therapeutic or diagnostic use.