Print…
VWR® EZ-Vision® In-gel Solution
Fornecedor: Avantor
VWR® EZ-Vision®
N391-0.5MLEA 78.4 EUR
N391-0.5ML N391-5MLDRP
VWR® EZ-Vision® In-gel Solution
Corantes Corantes para Electroforese
EZ-Vision in-gel solution is a non mutagenic and non toxic fluorescent DNA dye used as an in-gel or post-run stain to visualise DNA in agarose gels.
  • Immediate results, easy and convenient
  • Excellent replacement for the often used, but hazardous, ethidium bromide
  • Simply added to molten agarose prior to gel casting, allowing for instant visualisation of DNA bands upon UV exposure, as an in-gel preparation for electrophoresis