Print…

2-Chloro-6-(trifluoromethyl)nicotinic acid ≥98.0% (by titrimetric analysis)

Fornecedor: TCI EUROPE
Atenção

TCIAC2425-1GEA 90 EUR
TCIAC2425-1G
2-Chloro-6-(trifluoromethyl)nicotinic acid ≥98.0% (by titrimetric analysis)
2-Chloro-6-(trifluoromethyl)nicotinic acid
Fórmula: C₇H₃ClF₃NO₂
Peso Molecular: 225,56 g/mol
Número MDL : MFCD01862658
Número CAS: 280566-45-2
https://pt.vwr.com/stibo/web/std.lang.all/94/16/28939416.jpg

Encomendar

Especificações dos Resultados do Teste

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 113.0 to 120.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR