5-Acetylsalicylic acid ≥98.0% (by GC, titration analysis)
Fornecedor: TCI EUROPE
|
Atenção
|
TCIAA2378-5GEA
356
EUR
TCIAA2378-5G
5-Acetylsalicylic acid ≥98.0% (by GC, titration analysis)
5-Acetylsalicylic acid
Fórmula:
C₉H₈O₄ Peso Molecular: 180,15 g/mol |
Número MDL :
MFCD00013978 Número CAS: 13110-96-8 EINECS: 236-037-6 |
Especificações dos Resultados do Teste
Appearance | White to Light yellow powder to crystalline |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 216.0 to 220.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...