2,1,3-Benzothiadiazol-4-ylamine ≥98.0% (by GC, titration analysis)
Fornecedor: TCI EUROPE
|
Perigo
|
TCIAA1385-1GEA
53
EUR
TCIAA1385-1G
2,1,3-Benzothiadiazol-4-ylamine ≥98.0% (by GC, titration analysis)
2,1,3-Benzothiadiazol-4-ylamine
Fórmula:
C₆H₅N₃S Peso Molecular: 151,19 g/mol |
Número MDL :
MFCD00005810 Número CAS: 767-64-6 EINECS: 212-186-2 UN: 2811 ADR: 6.1,III |
Especificações dos Resultados do Teste
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 67.0 to 70.0 deg-C |
Solubility in Acetonitrile | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...