Print…
Reusable half masks against gas, vapour and dust, 8000 series
Fornecedor: Moldex
111-1178EA 26.6 EUR
MOLD8002
Reusable half masks against gas, vapour and dust, 8000 series
These versatile respirators can be used for gas/vapour protection, gas/vapour plus particulate protection, or only particulate protection dependent upon which combination of adapters and cartridges are used. The mask body is made from soft, durable TPE material.
  • Easy maintenance and cleaning without tools
  • Radial connector: New inhalation valve with every cartridge
  • Improved field of vision, lightweight and easy to fit
Certificações: EN 140:1998