Print…
Dropping bottle, LDPE
Fornecedor: Vitlab
VITL132293EA 83.6 EUR
VITL132293
Dropping bottle, LDPE
LDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
Narrow mouth transparent dropping bottle with dropper insert and red LDPE screw cap. This classic model, made from soft material with good restoring force.