Gostaria de reiniciar sessão?
PCR plates, 96-well, Armadillo™
Fornecedor: Thermo Scientific
Armadillo™
Rigid PC frame, thin-walled PP wells. Ultra-rigid 96-well PCR plates with rounded V-bottom wells designed for robotic applications.
  • Optically clear skirt aids visualisation of liquids and improves stacking capabilities in automated handling systems
  • Raised rim design improves seal integrity in automated heat sealing
  • With a rigid polycarbonate skirt, the ANSI/SBS footprint ensures complete compatibility with robotic systems for high throughput applications
  • Thermostable polycarbonate frame design allows warp-resistant thermal cycling and enhanced mechanical stability in robotic handling
  • Thin walled rounded V-bottom wells provides optimal heat transfer and maximum sample recovery
  • Low profile design for maximum heat transfer and minimal evaporation when utilising low volume samples (200 µl working volume)
  • Available with clear or white coloured wells ensuring the highest level of sensitivity for all PCR/qPCR reactions
Fornecimento: All plates can be provided with standard barcoding or barcoded to customer specifications for fully automated applications. Please contact your local VWR sales office for further information.
ENCOMENDA
Expandir tudo / Esconder tudo
Descrição Frame colour Well colour Volume do poço Und. Referência VWR Quantidade
Armadillo™ 96-well PCR plate transparente transparente 0,1 ml 25 731-0363
Armadillo™ 96-well PCR plate, barcoded transparente transparente 0,1 ml 25 731-0378
Armadillo™ 96-well PCR plate azul transparente 0,1 ml 25 731-0368
Armadillo™ 96-well PCR plate, barcoded azul transparente 0,1 ml 25 731-0379
Armadillo™ 96-well PCR plate verde transparente 0,1 ml 25 731-0369
Armadillo™ 96-well PCR plate, barcoded verde transparente 0,1 ml 25 731-0380
Armadillo™ 96-well PCR plate laranja transparente 0,1 ml 25 731-0370
Armadillo™ 96-well PCR plate, barcoded laranja transparente 0,1 ml 25 731-0381
Armadillo™ 96-well PCR plate vermelho transparente 0,1 ml 25 731-0371
Armadillo™ 96-well PCR plate, barcoded vermelho transparente 0,1 ml 25 731-0382
Armadillo™ 96-well PCR plate amarelo transparente 0,1 ml 25 731-0372
Armadillo™ 96-well PCR plate, barcoded amarelo transparente 0,1 ml 25 731-0383
Armadillo™ 96-well PCR plate, semi-skirted, low profile transparente transparente 0,1 ml 25 732-2793
Armadillo™ 96-well PCR plate, semi-skirted, low profile, barcoded transparente transparente 0,1 ml 25 732-2801
Armadillo™ 96-well PCR plate, semi-skirted, low profile azul transparente 0,1 ml 25 732-2794
Armadillo™ 96-well PCR plate, semi-skirted, low profile, barcoded azul transparente 0,1 ml 25 732-2802
Armadillo™ 96-well PCR plate, semi-skirted, low profile verde transparente 0,1 ml 25 732-2795
Armadillo™ 96-well PCR plate, semi-skirted, low profile, barcoded verde transparente 0,1 ml 25 732-2803
Armadillo™ 96-well PCR plate, semi-skirted, low profile laranja transparente 0,1 ml 25 732-2796
Armadillo™ 96-well PCR plate, semi-skirted, low profile, barcoded laranja transparente 0,1 ml 25 732-2804
Armadillo™ 96-well PCR plate, semi-skirted, low profile vermelho transparente 0,1 ml 25 732-2797
Armadillo™ 96-well PCR plate, semi-skirted, low profile, barcoded vermelho transparente 0,1 ml 25 732-2805
Armadillo™ 96-well PCR plate, semi-skirted, low profile amarelo transparente 0,1 ml 25 732-2798
Armadillo™ 96-well PCR plate, semi-skirted, low profile, barcoded amarelo transparente 0,1 ml 25 732-2806
Armadillo™ 96-well PCR plate, semi-skirted transparente transparente 0,2 ml 25 731-0644
Armadillo™ 96-well PCR plate, semi-skirted, barcoded transparente transparente 0,2 ml 25 731-0649
Armadillo™ 96-well PCR plate, semi-skirted azul transparente 0,2 ml 25 731-0655
Armadillo™ 96-well PCR plate, semi-skirted, barcoded azul transparente 0,2 ml 25 731-0652
Armadillo™ 96-well PCR plate, semi-skirted verde transparente 0,2 ml 25 731-0645
Armadillo™ 96-well PCR plate, semi-skirted, barcoded verde transparente 0,2 ml 25 731-0650
Armadillo™ 96-well PCR plate, semi-skirted laranja transparente 0,2 ml 25 731-0646
Armadillo™ 96-well PCR plate, semi-skirted, barcoded laranja transparente 0,2 ml 25 731-0651
Armadillo™ 96-well PCR plate, semi-skirted vermelho transparente 0,2 ml 25 731-0647
Armadillo™ 96-well PCR plate, semi-skirted, barcoded vermelho transparente 0,2 ml 25 731-0653
Armadillo™ 96-well PCR plate, semi-skirted amarelo transparente 0,2 ml 25 731-0648
Armadillo™ 96-well PCR plate, semi-skirted, barcoded amarelo transparente 0,2 ml 25 731-0654
Armadillo™ 96-well PCR plate transparente branco 0,1 ml 25 731-0364
Armadillo™ 96-well PCR plate, barcoded transparente branco 0,1 ml 25 731-0384
Armadillo™ 96-well PCR plate azul branco 0,1 ml 25 731-0373
Armadillo™ 96-well PCR plate, barcoded azul branco 0,1 ml 25 731-0385
Armadillo™ 96-well PCR plate verde branco 0,1 ml 25 731-0374
Armadillo™ 96-well PCR plate, barcoded verde branco 0,1 ml 25 731-0386
Armadillo™ 96-well PCR plate laranja branco 0,1 ml 25 731-0375
Armadillo™ 96-well PCR plate, barcoded laranja branco 0,1 ml 25 731-0387
Armadillo™ 96-well PCR plate vermelho branco 0,1 ml 25 731-0376
Armadillo™ 96-well PCR plate, barcoded vermelho branco 0,1 ml 25 731-0388
Armadillo™ 96-well PCR plate amarelo branco 0,1 ml 25 731-0377
Armadillo™ 96-well PCR plate, barcoded amarelo branco 0,1 ml 25 731-0389
Armadillo™ 96-well PCR plate, semi-skirted, low profile transparente branco 0,1 ml 25 732-2809
Armadillo™ 96-well PCR plate, semi-skirted, low profile, barcoded transparente branco 0,1 ml 25 732-2823
Armadillo™ 96-well PCR plate, semi-skirted, low profile azul branco 0,1 ml 25 732-2810
Armadillo™ 96-well PCR plate, semi-skirted, low profile, barcoded azul branco 0,1 ml 25 732-2824
Armadillo™ 96-well PCR plate, semi-skirted, low profile verde branco 0,1 ml 25 732-2811
Armadillo™ 96-well PCR plate, semi-skirted, low profile, barcoded verde branco 0,1 ml 25 732-2825
Armadillo™ 96-well PCR plate, semi-skirted, low profile laranja branco 0,1 ml 25 732-2812
Armadillo™ 96-well PCR plate, semi-skirted, low profile, barcoded laranja branco 0,1 ml 25 732-2826
Armadillo™ 96-well PCR plate, semi-skirted, low profile vermelho branco 0,1 ml 25 732-2813
Armadillo™ 96-well PCR plate, semi-skirted, low profile, barcoded vermelho branco 0,1 ml 25 732-2827
Armadillo™ 96-well PCR plate, semi-skirted, low profile amarelo branco 0,1 ml 25 732-2814
Armadillo™ 96-well PCR plate, semi-skirted, low profile, barcoded amarelo branco 0,1 ml 25 732-2828
Armadillo™ 96-well PCR plate, semi-skirted transparente branco 0,2 ml 25 732-2815
Armadillo™ 96-well PCR plate, semi-skirted, barcoded transparente branco 0,2 ml 25 732-2829
Armadillo™ 96-well PCR plate, semi-skirted azul branco 0,2 ml 25 732-2816
Armadillo™ 96-well PCR plate, semi-skirted, barcoded azul branco 0,2 ml 25 732-2830
Armadillo™ 96-well PCR plate, semi-skirted verde branco 0,2 ml 25 732-2817
Armadillo™ 96-well PCR plate, semi-skirted, barcoded verde branco 0,2 ml 25 732-2831
Armadillo™ 96-well PCR plate, semi-skirted laranja branco 0,2 ml 25 732-2818
Armadillo™ 96-well PCR plate, semi-skirted, barcoded laranja branco 0,2 ml 25 732-2832
Armadillo™ 96-well PCR plate, semi-skirted vermelho branco 0,2 ml 25 732-2819
Armadillo™ 96-well PCR plate, semi-skirted, barcoded vermelho branco 0,2 ml 25 732-2833
Armadillo™ 96-well PCR plate, semi-skirted amarelo branco 0,2 ml 25 732-2820
Armadillo™ 96-well PCR plate, semi-skirted, barcoded amarelo branco 0,2 ml 25 732-2834
Gerar códigos de barras Barcode Label Format: 
Criar página do catálogo em PDF
Detalhes
Selecione o artigo para o qual procura o Certificado
Indique o número do lote. Este campo é obrigatório.
Inserir detalhes do certificado
Número de Lote:
Choose from recent batch/lot numbers:
Ou
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Descrição Código de Artigo Disponibilidade Unidade O seu preço Preço por Qtd
Condições Gerais de Venda
Preços
Estas Condições Gerais de Venda anulam automaticamente todas as anteriores.
A VWR International – Material de Laboratório, Lda reserva-se o direito de alterar os preços sem aviso prévio.
Os preços dos produtos indicados serão os preços em vigor no momento da expedição, salvo cotação ou contrato válido.
Todos os preços estão sujeitos a ajustamentos dependentes de quantidades, embalagem especial, custo de matérias-primas ou de produção, expedição em condições especiais, etc.
Despesas de envio
Atendemos com a maior satisfação todos os pedidos, no entanto, reservamo-nos o direito de cobrar despesas de encomenda no valor de 9€ (no Continente) ou 28€ (nas Ilhas) e para encomendas inferiores a 195€ (valor sem IVA), valores de encomenda mínima, de 25€ e 50€ respetivamente.
Para encomendas efetuadas no nosso site, os valores anteriormente mencionados serão reduzidos para 5€ (despesas de encomenda no Continente), 15€ (encomenda mínima) e 47€ para as ilhas, respetivamente.
Relativamente a encomendas de produtos para os quais seja necessário o envio em condições especiais, será cobrado 25€ por despesas de transporte em condições de temperatura controlada e/ou 21,5€ por gastos de transporte especial (Avião). Esta cobrança será feita por cada encomenda.
Aos valores das despesas acima mencionados, acresce o IVA à taxa legal em vigor.
Caso o comprador reclame qualquer isenção, deverá apresentar um comprovativo de isenção válido.
Serviço
O Serviço de atendimento a clientes da VWR processará o seu pedido de uma forma mais rápida sempre que estiver munido das seguintes informações:
  • Nome da Empresa;
  • Número de cliente ou nº de contribuinte;
  • Endereço exato de entrega e de faturação (caso sejam diferentes), incluindo departamento, serviço, email, telefone, etc....);
  • Condições de descarga de mercadoria pesada
  • Códigos dos artigos encomendados de acordo com o catálogo;
  • Número de cotação (caso se aplique);
Para lhe proporcionarmos um melhor serviço e evitar erros, na confirmação por escrito de um pedido previamente efetuado por outra via, terá que, obrigatoriamente mencionar: "pedido previamente por telefone / fax / mail ou internet". A VWR não se responsabiliza por eventuais duplicações de encomendas nos casos em que o cliente não menciona a informação indicada. Qualquer encargo com a devolução do material aos nossos armazéns deverá ser suportado pelo cliente.
Logística e Distribuição
As encomendas serão entregues nos locais indicados na encomenda (de acordo com as condições particulares acordadas) e qualquer transporte a partir deste ponto é da inteira responsabilidade do cliente.
Procuramos entregar sempre os produtos o mais rápido possível. No entanto, todas as datas de entrega são aproximadas e a VWR International – Material de Laboratório, Lda. não poderá ser responsabilizada por qualquer perda ou dano decorrente de qualquer atraso na entrega por razões fora do seu controlo.
Solicitamos que verifique a quantidade e estado exterior das embalagens no ato de receção. Caso verifique qualquer anomalia nas mesmas (caixas danificadas, molhadas ou quantidade de caixas recebidas diferente da mencionada na guia de remessa) esta deverá ser assinalada no comprovativo de receção e reportada ao nosso Serviço de atendimento. Caso não seja feito qualquer registo da anomalia verificada no comprovativo de receção da mercadoria, a VWR não poderá assumir qualquer responsabilidade pelos danos de transporte, ficando todos os encargos a cargo do cliente.
Procedimento de Devoluções
A VWR International – Material de Laboratório, Lda., reserva-se o direito de não aceitar devoluções a não ser em casos excecionais. Todos os pedidos de devolução devem ser solicitados ao nosso Serviço de Atendimento de Clientes, no prazo de até 10 dias da data da guia de remessa. 
Só poderão ser aceites devoluções se os produtos se encontrarem nas suas embalagens originais e em perfeitas condições de revenda. A VWR não se responsabiliza por qualquer devolução que não seja feita mediante a apresentação da nossa guia de recolha por parte da transportadora.
As devoluções de material poderão estar sujeitas a um encargo de rearmazenamento no valor de 10% do valor de faturação (com um valor mínimo de 25€).
A VWR International – Material de Laboratório, Lda. não aceita devoluções de produtos especiais como por exemplo, de transporte e armazenamento em condições de temperatura controlada, produtos químicos ou biológicos de elevada perigosidade, etc. Não serão aceites devoluções efetuadas pelo cliente para os nossos escritórios.
Utilização dos produtos
Os produtos que a VWR fornece são destinados para uso de acordo com a documentação e especificações do fabricante e sob a responsabilidade do usuário. Contudo, para alguns produtos, tais como as susbtâncias ativas, a VWR, como distribuidor, não consegue assegurar o total cumprimento dos requistos legais (European guidelines for GMP medicinal products for human and veterinary use) e não pode vender os produtos em questão para essa finalidade.
Os produtos fornecidos pela VWR não podem ser usados como substâncias ativas para o fabrico de medicamentos de uso humano e veterinário. Para o uso dos produtos da VWR como produto farmacêutico, cosmético, aditivo alimentar e/ou agrícola, pesticida ou para uso doméstico, o cliente deve contatar o serviço local de atendimento da VWR.
O cliente que tenha intenção de utilizar os produtos da VWR como produto farmacêutico, cosmético, aditivo alimentar ou para outro uso para além do recomendado pela VWR devem realizar os seus próprios controlos internos para assegurar que os produtos se encontram em conformidade com a legislação local e diretrizes regulamentares (exemplo: Farmacopeia Europeia, etc...).
Em caso algum, a VWR será responsável se o cliente utilizar um produto numa aplicação diferente da recomendada.
É da responsabilidade do cliente assegurar a conformidade com a regulamentação aplicável (exemplo: a nível de Saúde, Segurança...) e tomar as precauções adequadas relativas ao armazenamento, manuseamento, venda e uso dos produtos.
Condições de Pagamento
Salvo condições específicas previamente acordadas, os preços dados são preços sem IVA com pagamento imediato no ato da encomenda. No caso de o pagamento ser efetuado de forma antecipada, será enviado para o correio electrónico indicado, uma confirmação de encomenda valorizada, sendo que a mesma só será expedida aquando do recebimento do pagamento.
A atribuição de crédito está condicionada à análise de alguns indicadores financeiros, podendo estes, vir a ser solicitados ao cliente.
Para avaliação de outras condições de crédito deverá ser efetuado um pedido por escrito para o email cdc.pt@vwr.com.
O não cumprimento dos prazos de pagamento previamente acordados, poderá ser sujeito a juros de mora, à taxa permitida por lei.
Formas de Pagamento
São aceites pela VWR International - Material de de Laboratório, Lda., os seguintes meios de pagamento: Débito directo, transferência bancária, cheque bancário, vale postal e numerário.
Considera-se como data de pagamento, o dia em que o valor é creditado na conta bancária da VWR International.
Jurisdição Competente
A Jurisdição para todos os litígios resultantes da distribuição de mercadoria e pagamentos é o Tribunal da Comarca de Oeiras.
Matriculada na Conservatóriado Registo Comercial de Cascais
Capital Social: : € 1.084.000
Contrib. Nº 503 842 770
Tel. Geral: + 351-21 3600 770
Fax: +0351-21 3600 798/9
Email: info.pt@vwr.com
https://www.vwr.com/
Print These Terms