Gostaria de reiniciar sessão?
CO₂ incubators with hot air sterilisation, CB 170 (Binder) 2018
Fornecedor: BINDER GMBH
390-1052 390-1053
CO₂ incubators with hot air sterilisation, CB 170 (Binder) 2018
The CB 170 CO2 incubator provides ideal, reproducible growth conditions for cell cultures. The deep-drawn stainless steel, obstruction free chamber without fan makes cleaning very easy. Uniform growth conditions are provided by the VENTAIR™ jacket system, this ensures accurate temperatures, homogeneous temperature distribution and the advanced Permadry™ humidity management with handy-sized water pan gives short recovery times after opening the door. DuoDoor allows the outer and inner glass door to be opened simultaneously if required.
  • Automatic sterilization cycle at 180 °C with sterilizable sensors
  • Intuitive touchscreen controller with time-segment and real-time programming .
  • USB port to download recorded internal data in open format
  • Enlarged 170 Liter inner volume keeping the same footprint as the CB 160 predecessor
  • FailSafe – Electronic error self-diagnostic system for safeguarding the cell cultures
  • Independent temperature safety device Class 3.1 (DIN 12880) with visual and audible temperature alarm protects the samples and the incubator
  • Stable pH values due to short CO2 recovery times with drift-free infrared CO2 measurement system
Units can be stacked using the stacking adaptor to save space, the lower stacking height of 1,80 m gives improved handling of the upper chamber, models have a tightly-fitted inner glass door made of safety glass and 2-point door lock.
Fornecimento: Supplied with 3 perforated stainless steel shelves and Binder test certificate.
Order Now
ENCOMENDA
ESPECIFICAÇÕES
ACESSÓRIOS
Inserir detalhes do certificado
Número de Lote:
Choose from recent batch/lot numbers:
Ou
Review & Compare Alternatives
Original Product:
Descrição Código de Artigo Disponibilidade Unidade O seu preço Preço por Qtd
Capital Social: : € 1.084.000
Contrib. Nº 503 842 770
Tel. Geral: + 351-21 3600 770
Fax: +0351-21 3600 798/9
Email: info.pt@vwr.com
https://www.vwr.com/
Print These Terms